Would you like to log in again?
Z-LEHD-Fmk
Supplier: Enzo Life Sciences
260-145-R100 260-145-R020
89146-966EA 258.76 USD InStock
89146-966 89146-964
Z-LEHD-Fmk
Proteins and Peptides
Caspase-9 inhibitor
Irreversible, cell permeable inhibitor of caspase-9.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty