Would you like to log in again?
Z-YVAD-FMK (Ready-to-Use), Enzo Life Sciences
Supplier: Enzo Life Sciences
260-154-R020 260-154-R100
89146-980EA 273.74 USD InStock
89146-980 89146-978
Z-YVAD-FMK (Ready-to-Use), Enzo Life Sciences
Proteins and Peptides
Inhibitor of caspase-1 and -4
Irreversible, cell permeable inhibitor of caspase-1. Inhibits also caspase-4.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty