Would you like to log in again?
AMT hydrochloride ≥98%
Supplier: Enzo Life Sciences
270-033-M050 270-033-M010
89147-094EA 142.78 USD InStock
89147-094 89147-096
AMT hydrochloride ≥98%
AMT hydrochloride
Potent and selective inhibitor of inducible nitric oxide synthase (iNOS/NOS II).
CAS Number: 21463-31-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/74/57/45347457.jpg
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty