Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid ≥98%

Supplier: Enzo Life Sciences
Synonyms: Naproxen(S)-2-(6-Methoxynaphthalen-2-yl)propanoic acid(S)-(+)-6-Methoxy-alpha-methyl-2-naphthaleneacetic Acid

270-102-G005
89147-206EA 61.1 USD InStock
89147-206
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid ≥98%
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid
COX inhibitor

Cyclooxygenase (COX) inhibitor. Widely used non-steroidal anti-inflammatory drug.

NSAID.


Formula: C₁₄H₁₄O₃
MW: 230.26 g/mol
MDL Number: MFCD00010500
CAS Number: 22204-53-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/74/76/45347476.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty