Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ratjadone A (synthetic)

Supplier: Enzo Life Sciences

270-346-C002
89147-664EA 256.65 USD InStock
89147-664
Ratjadone A (synthetic)
Ratjadone A
Represents a class of natural compounds, which inhibit proliferation in eukaryotes by blocking nuclear export. As potent as leptomycin B (Prod. No. ALX-380-100) and specific for G1/S checkpoint. Cytotoxic secondary metabolite (IC50=50pg/ml in mouse cell line L929) that arrests tumor cells in the G1 phase at remarkably low concentrations (50pg/ml in HeLa cell line KB3.1). Inhibits the binding between the nuclear export signal (NES) of proteins and the chromosome maintenance region protein (CRM1). Anticancer compound. Belongs to the family of orphan ligands which include polyketides like leptomycin B, callystatin A and other related compounds.


Formula: C₂₈H₄₀O₅
MW: 456.6 g/mol
CAS Number: 163564-92-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/75/27/45347527.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty