Would you like to log in again?
EB-47 dihydrochloride dihydrate
Supplier: Enzo Life Sciences
Synonyms: 1-Piperazineacetamide,4-[1-(6-amino-9H-purin-9-yl)-1-deoxy-β-D-ribofuranuron]-N-(2,3-dihydro-1H-isoindol-4-yl)-1-one 2HCl 2H2O
270-383-M005 270-383-M001
89147-754EA 285.57 USD InStock
89147-754 89147-752
EB-47 dihydrochloride dihydrate
EB-47 dihydrochloride dihydrate
PARP-1 inhibitor
Very potent and water soluble PARP-1 inhibitor (IC50=45nM, 100% inhibition at 200nM). Shows cytoprotective effects against oxidative damage in cells and in in vivo models of reperfusion injury and inflammation.
https://us-legacy.vwr.com/stibo/web/std.lang.all/75/47/45347547.jpg
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty