Would you like to log in again?
Indirubin-3’-(2,3-dihydroxypropyl)oximether
Supplier: Enzo Life Sciences
Synonyms: E804
270-425-M001
89147-868EA 181.09 USD InStock
89147-868
Indirubin-3’-(2,3-dihydroxypropyl)oximether
Indirubin-3’-(2,3-dihydroxypropyl)oximether
Src kinase inhibitor
Inhibitor of c-Src (IC50=0.43µM) blocking downstream STAT3 signalling. Induces apoptosis in human cancer cells.
https://us-legacy.vwr.com/stibo/web/std.lang.all/75/61/45347561.jpg
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty