Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Compound 15e ≥99% (by HPLC)

Supplier: Enzo Life Sciences
Synonyms: PI3K-α Inhibitor3-(4-Morpholinothieno[3,2-d]pyrimidin-2-yl)phenol

270-455-M005 270-455-M001
89147-952EA 320 USD InStock
89147-952 89147-950
Compound 15e ≥99% (by HPLC)
Compound 15e
PI3 kinase inhibitor

Potent and specific inhibitor of phosphoinositide 3-kinasep110alpha isoform (IC50=0.0020µM). p110beta isoform (IC50=0.016µM)p110gamma isoform (IC50=0.66µM)C2beta isoform (IC50=0.22µM). Inhibits proliferation of A375 melanoma cells (IC50=0.58µM). Inhibits proliferation of U87MG human glioblastoma cells (IC50=1.10.µM).Inhibits proliferation of A2780 human ovarian cells (IC50=0.27µM).


Formula: C₁₆H₁₅N₃O₂S
MW: 313.4 g/mol
CAS Number: 371943-05-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/75/79/45347579.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty