Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ouabain octahydrate ≥98% (by TLC)

Supplier: Enzo Life Sciences
Synonyms: Strophanthin G3-[(6-Deoxy-α-L-mannopyranosyl)oxy]-1,5,11α-14,19-penta-hydroxycard-20(22)-enolide

350-066-M100
89149-006EA 49.1 USD InStock
89149-006
Ouabain octahydrate ≥98% (by TLC)
Ouabain octahydrate
Na+/K+ ATPase inhibitor

A steroidal inhibitor of cardiac sodium pumps (Na+/K+ ATPase) which has been recognized as an endogenous adrenal hormone. In the CNS it can be used as a tool to induce in vivo excitotoxicity. Ouabain, as well as other endogenous digitalis-like compounds, can down-regulate the expression of 14-3-3 proteins in rat lens.


Formula: C₂₉H₄₄O₁₂.8H₂O
MW: 584.65 g/mol (anhy)
Storage Temperature: Refrigerator
MDL Number: MFCD00149240
CAS Number: 11018-89-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/76/60/45347660.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty