Would you like to log in again?
Hellebrin
Supplier: Enzo Life Sciences
350-105-M005
89149-074EA 303.31 USD InStock
89149-074
Hellebrin
Hellebrin
Na+/K+ ATPase inhibitor
Cardiotonic glycoside which presents a bufadienolide-steroid structure. Besides ouabain (Prod. No. ALX-350-066), from the class of cardenolides hellebrin is the second unique water soluble cardiotonic glycoside. Extremely potent inhibitor of Na+/K+-ATPase (sodium pump) blocking the active efflux of Na+ and reuptake of K+ of this membrane enzyme. Shows a positive inotropic effect by increasing the intracellular Ca2+ concentration. Strong immunosuppressor inhibiting T cell activity with much higher potency than cortisol or cyclosporin A (Prod. No. BML-A195). Induces caspase-dependent apoptosis.
Isolated from Helleborus sp.
CAS Number: 13289-18-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/76/71/45347671.jpg
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty