Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bakuchiol ≥97% (by HPLC)

Supplier: Enzo Life Sciences
Synonyms: (S,E)-4-(3,7-Dimethyl-3-vinylocta-1,6-dien-1-yl)phenol

350-144-M001
89149-164EA 58.12 USD InStock
89149-164
Bakuchiol ≥97% (by HPLC)
Bakuchiol
PTP1B inhibitor

Inhibitor of protein tyrosine phosphatase 1B (PTB1B). Antioxidant. Inhibitor of mitochondrial lipid peroxidation. Inhibitor of inducible nitric oxide synthase (iNOS; NOS II) expression. DNA polymerase inhibitor. Shows antimicrobial and cytotoxic activity.

Isolated from plant Psoralea corylifolia.


Formula: C₁₈H₂₄O
MW: 256.3826 g/mol
MDL Number: MFCD01707441
CAS Number: 10309-37-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/76/86/45347686.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty