Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nourseothricin sulfate (from Streptomyces noursei) ≥85%

Supplier: Enzo Life Sciences
Synonyms: Streptothricin sulfateclonNATStreptothricin sulphateNTC

380-025-M005 380-025-M001
89149-846EA 60 USD InStock
89149-846 89149-848
Nourseothricin sulfate (from Streptomyces noursei) ≥85%
Nourseothricin sulfate
Antibiotic

  • Mixture of streptothricin F (n=1) and D (n=3)

Active against both Gram-positive and Gram-negative bacteria and some fungi.

White to off-white powder

Soluble in water

Packaging: Shipping temperature: Ambient


Formula: C₃₁H₅₈N₁₂O₁₀·C₁₉H₃₄N₈O₈·H₂SO₄
MW: 1359.47 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00165013
CAS Number: 96736-11-7
https://us-legacy.vwr.com/stibo/web/std.lang.all/04/33/45390433.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty