Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-Amino-TEMPO, free radical) ≥97% (by HPLC)

Supplier: Enzo Life Sciences
Synonyms: 4-Amino-1-oxy-2,2,6,6-tetramethylpiperidine4-Amino-2,2,6,6-tetramethylpiperidine 1-Oxyl

400-019-M050
89150-600EA 48 USD InStock
89150-600
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-Amino-TEMPO, free radical) ≥97% (by HPLC)
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-Amino-TEMPO, free radical)
Free radical compound.

  • Formulation: Orange to red solid or liquid.


Formula: C₉H₁₉N₂O
MW: 171.26 g/mol
MDL Number: MFCD00006479
CAS Number: 14691-88-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/95/45390595.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty