Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium orthovanadate

Supplier: Enzo Life Sciences

400-032-G005 400-032-G025
89150-614EA 104 USD InStock
89150-614 89150-612
Sodium orthovanadate
Sodium orthovanadate
Alkaline phosphatase inhibitor

Potent inhibitor of alkaline phosphatase and tyrosine phosphates. Regarding decavanadate: across the pH range of 1-13, simple aqueous solutions of [VO4]3- ion involve a dozen or more ionic species, both monomeric and oligomeric, whose abundances depend strongly on pH and [VO4]3- concentration. At pH 3.75 a stable complex can be prepared (see below in Fohr et al.) which contains ten vanadium atoms and hence is a decavanadate ion. When diluted to pH 7.2, the decavanadate species has a half-life time of about 2 hours. Fohr et al., also provide directions for generating control solutions of mixed vanadate complexes in which decavanadate is absent and monomeric orthovanadate is present. Decavanadate inhibits inositol 1,4,5-trisphosphate- (IP3) induced calcium release from permeabilized endocrine cells and the binding of [3H]IP3 to its receptor in cerebellar and adrenal cortical membranes.


Formula: Na₃O₄V
MW: 183.91 g/mol
MDL Number: MFCD00003511
CAS Number: 13721-39-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/98/45390598.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty