Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,4-Butanediol dimethanesulfonate ≥98% DNA alkylating agent

Supplier: Enzo Life Sciences
Synonyms: Butane-1,4-diyl dimethanesulfonate1,4-Butanediol dimethanesulphonateButane-1,4-diyl dimethanesulphonate1,4-Butanediol dimethanesulfonate

400-048-G010
89150-656EA 64.4 USD InStock
89150-656
1,4-Butanediol dimethanesulfonate ≥98% DNA alkylating agent
Busulfan
Antineoplastic bifunctional, DNA alkylating agent that causes DNA damage by cross-linking DNA and proteins

Causes double-base lesions mainly at 5’-GA-3’ and, to a lesser extent, at 5’-GG-3’ sequences. Forms an intrastrand cross-link at the 5’-GA-3’ sequence, in addition to monoalkylation. Selectively induces normal human fibroblast senescence through Erk and p38 pathways.


Formula: C₆H₁₄O₆S₂
MW: 246.29 g/mol
Melting Pt: 114-118°C
MDL Number: MFCD00007562
CAS Number: 55-98-1
ADR: 6.1,I
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/07/45390607.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty