Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Oxo-2,2,6,6-tetramethylpiperidine-1-oxyl ≥98%, orange powder

Supplier: Enzo Life Sciences
Warning
Synonyms: 4-Oxo-2,2,6,6-tetramethylpiperidinooxy4-Oxo-TEMPOTEMPONE

430-079-M250 430-079-G001
89151-034EA 15.22 USD InStock
89151-034 89151-032
4-Oxo-2,2,6,6-tetramethylpiperidine-1-oxyl ≥98%, orange powder
4-Oxo-2,2,6,6-tetramethylpiperidine-1-oxyl
Spin trapping reagent.

Spin trap, useful for both in vitro and in vivo experiments.

Soluble in acetone, DMSO, diethylether, 100% ethanol or water.


Formula: C₉H₁₆NO₂
MW: 170.23 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00006187
CAS Number: 2896-70-0
UN: 3077
ADR: 9,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/63/45390663.jpg
Order Now
Specification Test Results
Purity (assay) ≥98%
Identity Determined by IR
Appearance Orange crystalline powder

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty