Would you like to log in again?
Human Recombinant BCMA:Fc
Supplier: Enzo Life Sciences
522-026-C050
89151-732EA 577.37 USD InStock
89151-732
Human Recombinant BCMA:Fc
Proteins and Peptides
Produced in HEK 293 cells. The extracellular domain of human BCMA (aa 2-54) is fused at the C-terminus to the Fc portion of human IgG1.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty