Would you like to log in again?
Mouse Recombinant TL1A (soluble)
Supplier: Enzo Life Sciences
522-105-C010
89151-882EA 600.5 USD InStock
89151-882
Mouse Recombinant TL1A (soluble)
Proteins and Peptides
Produced in HEK 293 cells. The extracellular domain of mouse TL1A (aa 94-252) is fused at the N-terminus to a linker peptide (8 aa) and a FLAG®-tag.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty