Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-(+)-2-Amino-4-phosphonobutyric acid ≥97% (by HPLC), white crystalline powder

Supplier: Enzo Life Sciences
Warning
Synonyms: L-AP4L(+)-2-Amino-4-phosphonobutanoic acid

550-026-M025 550-026-M005
89152-018EA 125 USD InStock
89152-018 89152-020
(S)-(+)-2-Amino-4-phosphonobutyric acid ≥97% (by HPLC), white crystalline powder
(S)-(+)-2-Amino-4-phosphonobutyric acid
mGluR agonist.

Selective agonist for group III mGluRs. NMDA agonist. Potent synaptic depressant.

Soluble in dilute aqueous base.


Formula: C₄H₁₀NO₅P
MW: 183.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00083244
CAS Number: 23052-81-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/07/91/45390791.jpg
Order Now
Specification Test Results
Purity (HPLC) ≥97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty