Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(+)-cis-Diltiazem hydrochloride ≥99% (by HPLC)

Supplier: Enzo Life Sciences
Synonyms: (2S,3S)-5-(2-(Dimethylamino)ethyl)-2-(4-methoxyphenyl)-4-oxo-2,3,4,5-tetrahydrobenzo[b][1,4]thiazepin-3-yl acetate hydrochloride(+)-cis-3-Acetoxy-5-(2-dimethylaminoethyl)-2,3-dihydro-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one Hydrochloride(2S,3S)-(+)-3-Acetoxy-2,3-dihydro-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one HydrochlorideDiltiazem Hydrochloride

550-214-G001
89152-356EA 56.7 USD InStock
89152-356
(+)-cis-Diltiazem hydrochloride ≥99% (by HPLC)
(+)-cis-Diltiazem hydrochloride
Inhibitor of L-type Ca2+ channels. Regulates Ca2+ release from intracellular stores in neutrophils.

  • Ca2+ channel inhibitor

White to off-white powder.

Soluble in water (50mg/ml), methanol or formic acid.


Formula: C₂₂H₂₆N₂O₄S·HCl
MW: 450.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00069252
CAS Number: 33286-22-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/08/20/45390820.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty