Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lipopolysaccharides (from Salmonella minnesota), TLRgrade™

Supplier: Enzo Life Sciences
Danger
Synonyms: Lipopolysaccharide

581-015-L002
89152-778EA 311.71 USD InStock
89152-778
Lipopolysaccharides (from Salmonella minnesota), TLRgrade™
Lipopolysaccharides
TLR4 activator.

Rough (R)-form LPS, isolated and purified from Salmonella minnesota R345 (Rb mutant) by a modification of the PCP extraction method, converted to the uniform sodium salt form.

Strong activator of Toll-like receptor 4 (TLR4). Does not activate TLR2 or other TLRs as determined with splenocytes and macrophages from TLR4 deficient mice. No further re-extraction required. Smooth (S)-form LPS are commonly the preferred choice for whole animal studies, whereas Rough (R)-form LPS are primarily used in cellular in vitro activation studies.


Order Now
Specification Test Results
Concentration 1 mg/ml
Formulation Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water
Purity Absence of detectable protein or DNA contaminants with agonistic TLR activity

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty