Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ABD-F [=4-(Aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole] ≥99% (by HPLC)

Supplier: Enzo Life Sciences
Synonyms: 4-(Aminosulphonyl)-7-fluoro-2,1,3-benzoxadiazole4-(Aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole7-Fluoro-2,1,3-Benzoxadiazole-4-sulfonamide7-Fluoro-2,1,3-Benzoxadiazole-4-sulphonamide7-Fluoro-2,1,3-Benzoxadiazole-4-Sulfonamide

620-029-M050
89152-864EA 565 USD InStock
89152-864
ABD-F [=4-(Aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole] ≥99% (by HPLC)
ABD-F [=4-(Aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole]
Thiol dye

Highly reactive fluorogenic reagent for labelling thiol compounds. Selectively labels protein cysteine residues without side chain modification or N-terminal blockage. Potent vasorelaxant.


Formula: C₆H₄FN₃O₃S
MW: 217.17 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00036687
CAS Number: 91366-65-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/08/71/45390871.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty