Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RUNX3 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
803-320-C100
89153-476EA 527.95 USD InStock
89153-476
Anti-RUNX3 Mouse Monoclonal Antibody
Antibodies
RUNX (Runt-related transcription factor) family members play important roles in normal developmental processes and carcinogenesis. Of the three known RUNX family members, the ubiquitously expressed RUNX3 is involved in neurogenesis, T cell differentiation, tumorigenesis and is a component of the TGF-β induced tumor suppressor pathway. In gastric cancer cells, RUNX3 induces cell cycle arrest and apoptosis through upregulation of Bim, a proapoptotic BH3-only protein. In addition, RUNX3 is frequently inactivated in gastric cancer cells.

  • For Immunohistochemistry, Western Blot

Recognizes human and mouse RUNX3.

Type: Primary
Antigen: RUNX3
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty