Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CEACAM8 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
805-088-C100
89155-388EA 705.93 USD InStock
89155-388
Anti-CEACAM8 Mouse Monoclonal Antibody
Antibodies
CEA-related cell adhesion molecule 8 (CEACAM8) belongs to the carcinoembryonic antigen (CEA) gene family. It encodes a glycosylphosphatidylinositol (GPI)-linked glycoprotein with a molecular mass of of 95kD and is expressed in cells of the granulocyte-lineage. It is expressed in neutrophils and eosinophils and is characterized as a granulocyte-specific activation antigen. Therefore CEACAM8 could serve as a marker for granulocyte activities. Like all members of the CEA family, it consists of a single N domain, with structural homology to the immunoglobulin variable domains, followed by two immunoglobulin constant-like A and B domains.

  • For ELISA, Flow Cytometry

Recognizes human CEACAM8. Does not cross-react with human CEACAM1, 3, 4, 5, 6, 7, or PSG1.

Type: Primary
Antigen: CEACAM8
Clonality: Monoclonal
Clone: 0
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty