Would you like to log in again?
PMN-Elastase (Human) ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
850-265-KI01
89156-068EA 928.17 USD InStock
89156-068
PMN-Elastase (Human) ELISA Kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of human PMN-elastase in cell culture supernatants, plasma and other body fluids.
  • Assay Time: 3.5 hours
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty