Would you like to log in again?
PTX3 (human) Detection Set [for ELISA application], Enzo Life Sciences
Supplier: Enzo Life Sciences
850-299-KI01
89156-106EA 1224.19 USD InStock
89156-106
PTX3 (human) Detection Set [for ELISA application], Enzo Life Sciences
Assays ELISAs
For the quantitative determination of PTX3 in human and bovine (weak) cell culture media, EDTA-plasma or serum (weak) samples.
Delivery information: Kit includes 1 vial (10 µg) PTX3 (human), 1 vial (50 µg) PTX3, pAb (Biotin), 1 vial (100 µg) PTX3, mAb (MNB4) for preparing 10 plates.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty