Would you like to log in again?
sHLA-G ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
850-309-KI01
89156-124EA 2034.3 USD InStock
89156-124
sHLA-G ELISA Kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of soluble human HLA-G (leukocyte antigen-G) in plasma, serum, cell culture supernatant and other body fluids
Cross-reacts strongly with bovine, dog, goat, monkey, rabbit and sheep HLA-G. Cross-reacts weakly with cat HLA-G.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty