Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Spectinomycin sulphate ≥98% (by HPLC)

Supplier: Enzo Life Sciences

BML-A281-0010
89156-368EA 97.1 USD InStock
89156-368
Spectinomycin sulphate ≥98% (by HPLC)
Spectinomycin sulphate
Antibiotic

  • White solid

Inhibits protein synthesis (elongation) by interfering with peptidyl tRNA translocation. Effective against Gram-negative and Gram-positive bacteria (Gonnococcus only). Mutation in rpsE (the gene for ribosomal protein S5) prevents binding of spectinomycin.

Soluble in water

Packaging: Shipping temperature: Ambient


Formula: C₁₄H₂₄N₂O₇·H₂SO₄
Storage Temperature: Freezer
MDL Number: MFCD00270185
CAS Number: 23312-56-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/34/38/45403438.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty