Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CK7 Mouse Monoclonal Antibody [clone: 4F5]

Supplier: Enzo Life Sciences
CA1139-0100
89156-764EA 272.31 USD InStock
89156-764
Anti-CK7 Mouse Monoclonal Antibody [clone: 4F5]
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunocytochemistry, Western Blot

Recognizes human, mouse, rat, pig cytokeratin proteins 7, 17 and 19 respectively. Detects bands of 54 kDa, 46 kDa and 40 kDa in Western Blot.

Type: Primary
Antigen: CK7
Clonality: Monoclonal
Clone: 4F5
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty