Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2',4'-Dichlorobenzamil HCl ≥95% (by HPLC)

Supplier: Enzo Life Sciences

BML-CA204-0005
89156-772EA 114.93 USD InStock
89156-772
2',4'-Dichlorobenzamil HCl ≥95% (by HPLC)
2',4'-Dichlorobenzamil HCl
Inhibits cyclic nucleotide-gated Ca2+ channels (IC50=38-50µM). Inhibits plasmalemmal Na+/Ca2+ and Na+/H+ exchange (IC50=10µM). Blocks caffeine-induced current (by blocking Na+/Ca2+ exchange) at 50-100µM. Nonselective cation channel blocker (25µM).

  • Ca2+ channel blocker

Yellow solid.

Soluble in DMSO (25 mg/ml), Ethanol (25 mg/ml) or DMF (25 mg/ml).


Formula: C₁₃H₁₂Cl₃N₇O·HCl
MW: 425.1 g/mol
Storage Temperature: Ambient
CAS Number: 2088-58-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/34/71/45403471.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty