Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Indirubin-3'-oxime ≥98%

Supplier: Enzo Life Sciences

BML-CC207-0005 BML-CC207-0001
89156-906EA 109.7 USD InStock
89156-906 89156-908
Indirubin-3'-oxime ≥98%
Indirubin-3'-oxime
A potent inhibitor of glycogen synthase kinase-3β (IC50=22 nM). Also inhibits CDK1 (p34cdc2; IC50=180 nM) and CDK5 (IC50=100 nM). It reversibly arrests asynchronous HBL-100 cells at G2. Induces apoptosis in the mammary carcinoma cell line MCF-7 (10 µM).

  • Inhibitor of CDK and GSK

Dark red solid.

Soluble in DMSO (6mg/ml) and ethanol (4mg/ml).


Formula: C₁₆H₁₁N₃O₂
MW: 277.283 g/mol
Storage Temperature: Freezer
MDL Number: MFCD02683594
CAS Number: 160807-49-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/35/02/45403502.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty