Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dibutyryl-cGMP ≥96%

Supplier: Enzo Life Sciences

BML-CN215-0050 BML-CN215-0010
89157-040EA 133.71 USD InStock
89157-040 89157-042
Dibutyryl-cGMP ≥96%
Dibutyryl-cGMP
A cell-permeable cGMP analog which preferentially activates cGMP-dependent protein kinase. Inhibits thrombin stimulated arachidonic acid release in human platelets. Cell permeable.

  • PKG activator

White solid.

Soluble in water (100mg/ml). Also soluble in 100% ethanol, DMSO, dimethyl formamide or methanol.


Formula: C₁₈H₂₃N₅O₉P·Na
MW: 484.4 · 23.0 g/mol
Storage Temperature: Freezer
CAS Number: 51116-00-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty