Would you like to log in again?
Diphenyleneiodonium chloride ≥98%
Supplier: Enzo Life Sciences
Synonyms: Diphenyleneiodonium Chloride
BML-CN240-0010
89157-048EA 79.6 USD InStock
89157-048
Diphenyleneiodonium chloride ≥98%
Diphenyleneiodonium chloride
NADPH oxidase inhibitor
Binds strongly to flavoproteins and is thus a powerful and specific inhibitor of several important enzymes, including nitric oxide synthase (NOS), NADPH-ubiquinone oxidoreductase, NADPH oxidases and NADPH cytochrome P450 oxidoreductase. Nitric oxide synthase, which shows significant homology with cytochrome P450 reductase, has been shown to be irreversibly inhibited by this compound in cultured peritoneal macrophages (IC50=30nM).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty