Would you like to log in again?
Blocking Peptide For Dishevelled-2 pAb (Prod. No. BmL-DA4270) Blocking Peptide
Supplier: Enzo Life Sciences
DP9427-0100
89157-384EA 82.63 USD InStock
89157-384
Blocking Peptide For Dishevelled-2 pAb (Prod. No. BmL-DA4270) Blocking Peptide
Proteins and Peptides
Sequence derived from human/mouse dishevelled-2 [Cys+600-614]. Used to generate the rabbit polyclonal antiserum to dishevelled-2 (BML-DA4270). The peptide is suitable for use in negative control (antibody blocking) experiments in Western blotting and immunohistochemistry.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty