Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-(-)-2-Amino-5-phosphonopentanoic acid ≥99% (by HPLC), white to off-white solid

Supplier: Enzo Life Sciences
Warning
Synonyms: (R)-2-Amino-5-phosphonopentanoic acidD-2-Amino-5-phosphonovaleric acidD-AP5

BML-EA104-0005
89157-416EA 133 USD InStock
89157-416
(R)-(-)-2-Amino-5-phosphonopentanoic acid ≥99% (by HPLC), white to off-white solid
(R)-(-)-2-Amino-5-phosphonopentanoic acid
NMDA receptor antagonist

Active form of AP5. Potent NMDA receptor antagonist.

Soluble in water (acidify for better solubility).

Caution: Store, as supplied, at room temperature for up to 1 year. Store solutions at -20 °C for up to 3 months.


Formula: C₅H₁₂NO₅P
MW: 197.127 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078839
CAS Number: 79055-68-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/36/00/45403600.jpg
Order Now
Specification Test Results
Purity ≥99% (HPLC)
Identity Determined by H-NMR

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty