Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Esculetin ≥98%

Supplier: Enzo Life Sciences
Synonyms: 6,7-Dihydroxy-2H-chromen-2-one6,7-Dihydroxycoumarin

BML-EI100-0001
89157-550EA 50.2 USD InStock
89157-550
Esculetin ≥98%
Esculetin
Lipoxygenase inhibitor

Inhibitor of 5-lipoxygenase (ID50=4 µM) and 12-lipoxygenase (ID50=2.5 µM). Does not inhibit cyclooxygenase.


Formula: C₉H₆O₄
MW: 178.14 g/mol
MDL Number: MFCD00006874
CAS Number: 305-01-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/36/11/45403611.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty