Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Manoalide ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-EI177-0001
89157-718EA 265.01 USD InStock
89157-718
Manoalide ≥98% (by TLC)
Manoalide
Phospholipase inhibitor

Marine sesterterpenoid. Irreversibly inhibits human synovial phospholipase A2 (PLA2, IC50=0.02-0.2µM depending on substrate) as well as bee (IC50=0.05µM) and cobra venom (IC50=2µM) PLA2 by covalently modifying lysine residues. Inhibits human recombinant sPLA2 (IC50=4.7µM) selectively over cPLA2. Also inhibits phospholipase C (PLC, IC50=1.5µM) and Ca2+ channels. Inhibits superoxide generation in polymorphonuclear leukocytes. Inhibits the release of arachidonate in cultured mouse peritoneal macrophages and in a mouse in vivo model. Has anti-inflammatory and analgesic properties. Shows strong quorum sensing antagonism.


Formula: C₂₅H₃₆O₅
MW: 416.558 g/mol
MDL Number: MFCD00153826
CAS Number: 75088-80-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/36/74/45403674.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty