Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tyrphostin AG 183 ≥99%

Supplier: Enzo Life Sciences
Synonyms: Tyrphostin A513-Amino-2,4-dicyano-5-(3',4',5'-trihydroxyphenyl)penta-2,4-dienonitrile

BML-EI189-0005
89157-738EA 112.84 USD InStock
89157-738
Tyrphostin AG 183 ≥99%
Tyrphostin AG 183
A potent inhibitor of EGF receptor kinase activity (IC50=0.8 µM). Inhibition is mixed competitive with ATP and substrate.

  • EGFR inhibitor

Brown solid.

Slightly soluble in water.


Formula: C₁₃H₈N₄O₃
MW: 268.2 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00133903
CAS Number: 122520-90-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/36/80/45403680.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty