Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calphostin C ≥99% (by TLC)

Supplier: Enzo Life Sciences

BML-EI198-1000 BML-EI198-0100
89157-766EA 159 USD InStock
89157-766 89157-768
Calphostin C ≥99% (by TLC)
Calphostin C
PKC inhibitor

Potent and selective inhibitor of the protein kinase C (PKC). Inhibition is via interaction with the regulatory DAG binding site and phorbol ester binding site. Inhibition of PKC has been found to be light dependent. At higher concentrations inhibits myosin light chain kinase, cAMP-dependent protein kinase, protein kinase G, pp60v-src protein tyrosine kinase and DAG kinase. It also inhibits Phospholipase D1 and D2. Induces apoptotic DNA fragmentation and cell death. Kills breast cancer cells. Has antiviral potential. Inhibits cardiac L-type Ca2+ channels. Cell permeable.


Formula: C₄₄H₃₈O₁₄
MDL Number: MFCD00133155
CAS Number: 121263-19-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/36/88/45403688.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty