Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Aminobenzotriazole ≥99%

Supplier: Enzo Life Sciences
Synonyms: 1H-Benzo[d][1,2,3]triazol-1-amine

BML-EI259-0100
89157-898EA 175.13 USD InStock
89157-898
1-Aminobenzotriazole ≥99%
1-Aminobenzotriazole
Cytochrome P450 inhibitor

Suicide substrate for cytochrome P-450. Inhibits oxidative drug metabolism in vivo. Blocks omega-oxidation of arachidonic acid but not epoxidation.


Formula: C₆H₆N₄
MW: 134.14 g/mol
MDL Number: MFCD00132902
CAS Number: 1614-12-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/37/17/45403717.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty