Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

7-Ethoxyresorufin ≥98%

Supplier: Enzo Life Sciences
Synonyms: EthoxyresorufinResorufin ethyl ether

BML-EI262-0010
89157-910EA 295 USD InStock
89157-910
7-Ethoxyresorufin ≥98%
7-Ethoxyresorufin
Cytochrome P450 substrate

Fluorimetric substrate for cytochrome P450 IA1. Inhibits vasorelaxant responses to acetylcholine.


Formula: C₁₄H₁₁NO₃
MW: 241.246 g/mol
MDL Number: MFCD00037661
CAS Number: 5725-91-7
https://us-legacy.vwr.com/stibo/web/std.lang.all/37/19/45403719.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty