Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ikarugamycin ≥95% (by HPLC, TLC)

Supplier: Enzo Life Sciences

BML-EI313-0500
89158-048EA 288.7 USD InStock
89158-048
Ikarugamycin ≥95% (by HPLC, TLC)
Ikarugamycin
Ikarugamycin inhibits the uptake of oxidized low-density lipoprotein in mouse macrophages

  • White to off-white crystalline solid

It blocks PMA and Nef-mediated cell surface CD4 down-regulation. Ikarugamycin is emerging as a general inhibitor of clathrin-coated pit-mediated endocytosis and appears to be a useful agent for studying the process of endocytosis.

Soluble in DMFA, DMSO or THF

Packaging: Shipping temperature: Ambient


Formula: C₂₉H₃₈N₂O₄
MW: 478.6 g/mol
Storage Temperature: Freezer
CAS Number: 36531-78-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/37/62/45403762.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty