Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Exo1 ≥98%

Supplier: Enzo Life Sciences
Synonyms: 4-Fluoro-Bz-2-Abz-OMe

BML-EI322-0010
89158-070EA 96.43 USD InStock
89158-070
Exo1 ≥98%
Exo1
ADP-ribosylation factor inhibitor

Exo1 inhibits traffic emanating from the endoplasmic reticulum by inducing rapid collapse of the Golgi to the endoplasmic reticulum1. Like Brefeldin A, Exo1 induces the rapid release of ARF1 from Golgi membranes, but evidence suggests that Exo1 acts by a different mechanism. Exo1 does not induce the ADP-ribosylation of CtBP/Bars50, and does not interfere with the activity of guanine nucleotide exchange factors specific for Golgi-based ARFs, allowing Exo1 to be used as a tool for differentiating the fatty acid exchange activity of Bars50 from ARF1 activity.


Formula: C₁₅H₁₂FNO₃
MW: 273.263 g/mol
CAS Number: 75541-83-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/37/71/45403771.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty