Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Arachidonylglycine ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-FA029-0005 BML-FA029-0025
89158-402EA 380 USD InStock
89158-402 89158-400
N-Arachidonylglycine ≥98% (by TLC)
N-Arachidonylglycine
FAAH inhibitor

Endogenous anandamide-like compound. Lacks affinity for CB1 receptors (Ki>10µM), TRPV1 (EC50>10µM) and anandamide uptake (IC50>50µM), but inhibits fatty acid amide hydrolase (FAAH) (IC50=8.5µM-50µM, depending on cell type and species). Displays anti-inflammatory and analgesic activity. Identified as an insulin secretagogue in a primary beta-cell-based functional assay. Represents a new class of "lipoamino acids".


Formula: C₂₂H₃₅NO₃
MW: 361.526 g/mol
MDL Number: MFCD03791313
CAS Number: 179113-91-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/38/51/45403851.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty