Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GLP1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
GA1176-0100
89158-690EA 487.02 USD InStock
89158-690
Anti-GLP1 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunohistochemistry

Recognizes mammalian GLP1.

Type: Primary
Antigen: GLP-1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mammals

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty