Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tanshinone IIA ≥98% (by TLC)

Supplier: Enzo Life Sciences
Synonyms: Dan Shen Ketone1,6,6-Trimethyl-6,7,8,9-tetrahydrophenanthro[1,2-b]furan-10,11-dione

BML-GR336-0005 BML-GR336-0025
89158-964EA 487.54 USD InStock
89158-964 89158-962
Tanshinone IIA ≥98% (by TLC)
Tanshinone IIA
AP-1 inhibitor

Tanshinone IIA is a diterpenoid naphthoquinone found in the traditional Chinese medicine Tanshen (Salvia sp). It inhibits AP-1 activity by suppressing jun-fos-DNA complex formation (IC50=0.22 µM) and induces apoptosis in a variety of cell lines


Formula: C₁₉H₁₈O₃
MW: 294.35 g/mol
MDL Number: MFCD00238692
CAS Number: 568-72-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/12/64/45401264.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty