Synonyms:
N-Palmitoyl-L-serine phosphoric acid
BML-LP101-0001
89160-536EA
134.75
USD
InStock
89160-536
L-NASPA ≥98% (by TLC)
L-NASPA
L-NASPA is an antagonist of LPA-induced platelet aggregation and chloride conductance in Xenopus oocytes. In human breast carcinoma MDA-MB231 cells it is a potent LPA agonist.
Soluble in DMSO (10 mg/ml)or ethanol (10 mg/ml).
Synonym: N-Palmitoyl-L-serine phosphoric acid