Would you like to log in again?
Ac-Leu-Leu-Nle-CHO
Supplier: Enzo Life Sciences
BML-P120-0025 BML-P120-0005
89160-954EA 498.91 USD InStock
89160-954 89160-952
Ac-Leu-Leu-Nle-CHO
Proteins and Peptides
Cysteine protease inhibitor
Cell-permeable, peptide aldehyde inhibitor of calpain I (Ki=190nM), calpain II (Ki=150nM), cathepsin L (Ki=0.5nM) and other neutral cysteine proteases. Inhibits cell cycle progression at G1/S and metaphase/anaphase in CHO cells by inhibiting cyclin B degradation. Also stimulates HMG-CoA synthase transcription by inhibiting degradation of active SREBP-1 (sterol regulatory element-binding protein 1). Protects against neuronal damage caused by hypoxia and ischemia. Inhibits apoptosis in thymocytes and metamyelocytes. Also prevents nitric oxide production by activated macrophages by interfering with the transcription of inducible nitric oxide synthase (iNOS; NOS II). Inhibits proteolytic degradation of IκBα and IκBß in RAW macrophages induced with LPS. It also prolong association of MHC class I molecules with the transporters associated with antigen processing.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty