Would you like to log in again?
Ikb Kinase Substrate,Biotin
Supplier: Enzo Life Sciences
BML-P148-0001
89161-010EA 211.35 USD InStock
89161-010
Ikb Kinase Substrate,Biotin
Proteins and Peptides
This peptide sequence, RHDSGLDSMKD, is derived from the area surrounding serine 32 and serine 36 of IκBα, which is phosphorylated by IκB Kinase (IKK). The biotin allows the peptide to be used in IKK kinase assays with streptavidin-bound membranes.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty