Would you like to log in again?
mLCK
Supplier: Enzo Life Sciences
BML-P209-0001
89161-082EA 167.48 USD InStock
89161-082
mLCK
Proteins and Peptides
This sequence, KRRWKKNFIAV, is from skeletal muscle myosin light chain kinase (MLCK) (342-352) pseudosubstrate site. Inhibits MLCK (Ki =1 µM with MLC peptide substrate). Possesses only slight calmodulin binding activity.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty